Learn More
R&D Systems™ High-Throughput Glutathione Peroxidase Assay Kit
Product Code. 30001328
Change view
Click to view available options
Quantity:
480 Tests
Unit Size:
480 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001328 480 Tests 480 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30001328 Supplier R&D Systems™ Supplier No. 7512100K
  • ONLINE EXCLUSIVE PRICE
    783.00€
    740.25€ / 480 tests
    Save 42.75€
    5% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Includes
Glutathione Peroxidase, GSH and NADPH, Glutathione Reductase, Cumene Hydroperoxide, Buffer, 5 x 96-well Plates
Spectrophotometric assay for the quantitation of glutathione peroxidase in cells, erythrocytes, blood, and tissues
Glutathione peroxidase is a tetramer of four identical subunits with MW 84,000. It is found in mammalian cells and helps prevent lipid peroxidation of cell membranes by consuming peroxide in the cell.
Sufficient reagents for 480 Tests-96 well Format or 100 Tests-cuvette Format
Rate of decrease in the A340 is directly proportional to the glutathione peroxidase activity in the sample
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The product requires storage at -20 to -70° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Quantity 480 Tests
Product Line R&D Systems