Learn More
Macherey-Nagel™ High Temperature Septa
Click to view available options
Diameter (Metric) Outer:
9 mm
10 mm
11 mm
12 mm
17 mm
Unit Size:
Pack of 50
Specifications
Specifications
| Color | Red |
| Diameter (Metric) Outer | 11 mm |
| Material | Red Non-bleeding Silicone |
| Septum | High Temperature Septa |
| Temperature Range (Metric) | 320°C |
| Quantity | 50 Pk. |
| Thickness (Metric) | 3 mm |