Learn More
Applied Biosystems™ High Resolution Melt (HRM) Plug-in for DA3 Software
Shop All Applied Biosystems ProductsSpecifications
Specifications
| For Use With (Equipment) | QuantStudio 1, QuantStudio 3 and 5, QuantStudio 6 and 7 Pro, StepOne and StepOnePlus Real Time PCR Systems, and The 7500 Fast Real Time PCR System |
| Software Compatibility | Diomni Design and Analysis v3.1 Desktop Application, and DA2 Software |
| Quantity | 1 License |