Learn More
Cytiva High Purity rNTP Set
Product Code. 10655355 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4 x 25 μmol
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10655355 4 x 25 μmol 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10655355 Supplier Cytiva Supplier No. 27202501
  • ONLINE EXCLUSIVE PRICE
    284.00€
    259.62€ / 1 sets
    Save 24.38€
    8% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Use for in vitro transcription or for reactions requiring rNTPs of high purity. Cytiva Life Sciences™ High Purity rNTP Set is easy-to-use and offers time-saving solutions.
  • Each lot is tested for ribonuclease contamination to ensure success in your application
  • High purity: >98% triphosphate
  • In a convenient set containing ATP, CTP, GTP, and UTP in separate tubes
TRUSTED_SUSTAINABILITY
Specifications
Format Liquid
Description Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
pH 7.5
Product Type rNTP Set, 100 mM Solutions (ATP, CTP, GTP, UTP)
Concentration 100 mM
Purity ≥98% triphosphate
Quantity 4 x 25 μmol
For Use With (Application) in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling