Learn More
Cytiva High Purity dNTPs
Used for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling. Cytiva High Purity dNTPs are tested functionally in long PCR* to be PCR-quantified for high-quality sequencing.
469.00€ - 553.00€
Specifications
Format | Solid |
---|---|
Content And Storage | -20°C |
Purity | ≥98% triphosphate |
Quantity | 250 mg |
For Use With (Application) | Amplification, Dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11505115
|
Cytiva
27-1850-04 |
dATP |
469.00€
250mg |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
11525115
|
Cytiva
27-1870-04 |
dGTP |
553.00€
250mg |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
11515115
|
Cytiva
27-1860-04 |
dCTP |
553.00€
250mg |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
11535115
|
Cytiva
27-1880-04 |
dTTP |
553.00€
250mg |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
Description
- High purity: >98% triphosphate, as confirmed by chromatographic analysis and 31P nuclear magnetic resonance (NMR) spectroscopy
- Supplied as solids and also available in solutions
Specifications
Solid | |
≥98% triphosphate | |
Amplification, Dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling |
-20°C | |
250 mg |