missing translation for 'onlineSavingsMsg'
Learn More

ANSELL™ High Performance Reusable Food-approved AlphaTec™ Gloves

Product Code. 17993572 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 17993572

Brand: ANSELL™ 58010110

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Durable yet ergonomic 0.28 mm nitrile glove.

  • Chemical glove with all day comfort which strikes a fantastic balance between value and durability
  • 11 mil or 0.28 mm thickness allows dexterity and tactility without a loss of durability
  • NBR offering protection against light acids, greases, oils and liquids
  • Engineered fit to allow optimum balance of fit and flexibility both on its own or as an over glove
  • Long cuff extended protection to the forearm
  • Silicone and latex free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) ANSI ABR 4|EN 388:2016 +A1:2018|EN 421:2010|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Food Contact
Chlorinated Yes
Applications Industrial
Color Blue
Finger Thickness (English) 11 mil
Finger Thickness (Metric) 0.28 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 11 mil
Palm Thickness (Metric) 0.28 mm
Size 11
Thickness (English) 11 mil
Thickness (Metric) 0.28 mm
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Raised Diamond
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.