Learn More
ANSELL™ High Performance Reusable Food-approved AlphaTec™ Gloves
Product Code. 17983572 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17983572 8 12 pairs
17923542 9 12 pairs
17903542 7 12 pairs
17973572 10 12 pairs
17993572 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17983572 Supplier ANSELL™ Supplier No. 58010080
Only null left
Add to basket
Add to basket
Durable yet ergonomic 0.28 mm nitrile glove.
  • Chemical glove with all day comfort which strikes a fantastic balance between value and durability
  • 11 mil or 0.28 mm thickness allows dexterity and tactility without a loss of durability
  • NBR offering protection against light acids, greases, oils and liquids
  • Engineered fit to allow optimum balance of fit and flexibility both on its own or as an over glove
  • Long cuff extended protection to the forearm
  • Silicone and latex free
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) ANSI ABR 4|EN 388:2016 +A1:2018|EN 421:2010|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Food Contact
Chlorinated Yes
Applications Industrial
Color Blue
Finger Thickness (English) 11 mil
Finger Thickness (Metric) 0.28 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 11 mil
Palm Thickness (Metric) 0.28 mm
Product Type Chemical-resistant Gloves
Size 8
Thickness (English) 11 mil
Thickness (Metric) 0.28 mm
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Raised Diamond
Lining Unlined