Learn More
DWK Life Sciences Wheaton™ Crimping Jaw Set for High Performance Crimping Tool
Product Code. p-6830049 Shop All Wheaton Science Products Products
Click to view available options
Size:
20 mm
For Use With (Application):
Crimping
Unit Size:
Each
Product Code. 15778979
Brand: DWK Life Sciences W225771
Edge
Interchangeable jaws allow users to choose a jaw that best fits their needs.
Available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) High performance crimping tool
Size 20 mm
For Use With (Application) Crimping