missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Duplex, Zeron 100, S32760, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, Duplex, Zeron 100, S32760, Ø38x20mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Phosphorus/Niobium/Cobalt/Vanadium/Nitrogen/Silicon/Copper/Manganese/Tungsten/Molybdenum/Nickel/Chromium |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon 0.021 %/Phosphorus 0.025 %/Niobium 0.026 %/Cobalt 0.044 %/Vanadium 0.058 %/Nitrogen 0.23 %/Silicon 0.29 %/Copper 0.59 %/Manganese 0.59 %/Tungsten 0.59 %/Molybdenum 3.5 %/Nickel 7.48 %/Chromium 25.5 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction