Learn More
UVP High Intensity Longwave UV Lamp
Click to view available options
Wattage:
100 W
Unit Size:
Each
Specifications
Specifications
| Voltage | 230 V |
| Wattage | 100 W |
| Electrical Requirements | 100 W, 230 V, 60 Hz |
| Lighting Intensity | 21,700/8900 μW/cm2 |
| Product Type | High Intensity Longwave UV Lamp |