Learn More
Heidolph™ High Hood
Shop All Heidolph Instruments Products
Click to view available options
For Use With (Equipment):
Incubator
Unit Size:
Each
Description
This hood has a height of 267 mm and is therefore ideally suited for Erlenmeyer flasks up to 500 mL
Specifications
Specifications
| Voltage | 115/230 V |
| Product Type | High Hood |
| Dimensions (L x W x H) | 394 x 546 x 267 mm |
| For Use With (Equipment) | Incubator |