Learn More
Mutexil™ High Filtration Hygiene Face Mask
Product Code. 11745453
Change view
Click to view available options
:
50/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11745453 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11745453 Supplier Mutexil™ Supplier No. 3000017
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Nonwoven mask fits behind ears and adjusts to bridge of nose.
  • Round elastic, latex free
  • Nonwoven
  • 3-ply
  • Adjustable nose bridge
Specifications
Product Type Face Mask
Color Blue
Certifications/Compliance (Europe) EN 14683, RII
Construction 3-ply, non-woven
Description High filtration
Includes Round elastic, nose bridge
Latex Free Yes
Packaging Quantity 50/Pk.