Learn More
Taylor Wharton™ HC Series Cryogenic Storage Dewar
Product Code. 11708519
Click to view available options
Capacity (Metric):
20 L
35 L
Unit Size:
Each
Product Code. 11708519
Brand: Taylor Wharton™ HC20
Only null left
Add to basket
Add to basket
The HC series refrigerators are designed to provide greater storage capacity while the compact design makes it the ideal choice where space is limited. They are primarily utilised to store material in vials, or for applications involving larger quantities of straws.
  • Easy operation, lightweight one-piece snap-on lid and neck tube assures tight closure and easy access
  • Convenient larger handles
  • Advanced multi-layer insulation, latest vacuum technology systems and other premium materials provide leading industry performance
  • Engineering utilised finite element analysis, with specific testing to create durable aluminium dewars
Specifications
Description 20 L Cryogenic Dewar, holds vials or straws
Diameter (Metric) 396 mm
Model HC20
Static Evaporation Rate 0.23 L/Day
Static Holding Time 87 Days
Capacity (Metric) 20 L
Material Aluminum
For Use With (Application) Long term storage of biological material at -196°C
Height (English) Exterior 24.2 in.
Height (Metric) Exterior 615 mm
Weight (Metric) 12 kg (empty)