Learn More
Labconco High Capacity Micro and 12 x 32 Rotor - CentriVap Pro
GREENER_CHOICE
Product Code. 30533413 Shop All Labconco Corporation Products
Click to view available options
Unit Size:
Each
Material (Coating):
PTFE
Product Code. 30533413
Brand: Labconco 7555500
Rotors for use with Labconco™ CentriVap™ Pro Vacuum Concentrators. Stainless steel and PTFE coated options available.
  • Compatible with CentriVap Pro Vacuum Concentrators
  • Stackable design
  • Stainless steel material\
  • Optional PTFE coated
Specifications
Material Stainless Steel
Type Rotor
Rotor System CentriVap Pro
Height (English) 1.8 in.
Height (Metric) 4.57 cm
Diameter (English) 11.4 in.
Diameter (Metric) 28.96 cm