Learn More
Leica Microsystems Hi-Plan Objective
Product Code. 11965018
Change view
Click to view available options
Description:
100X / 1.25
10X / 0.22
40X / 0.65
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
11965018 100X / 1.25 Each
11945018 10X / 0.22 Each
11955018 40X / 0.65 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11965018 Supplier Leica Microsystems Supplier No. 11506238
Estimated Shipment: 15-10-2026
The Leica DM1000 is the ideal instrument for biological, medical and clinical laboratories. The instrument can be configured to meet specific requirements by choosing from a selection of observation tubes, ergonomic modules, objectives and adjustable controls. Please note a complete system comprises the main microscope unit, an observation tube and either the brightfield or phase contrast outfit. Fluorescence outfits also require the brightfield or phase contrast outfit to be ordered.
TRUSTED_SUSTAINABILITY
Specifications
Description 100X / 1.25