Learn More
CPC (Colder) HFC39 Quick-Disconnect Fittings, Hose Barb Insert, Polysulfone
Product Code. 30054082
Click to view available options
Diameter (English) Inner:
1/2 in.
1/4 in.
3/8 in.
Product Type:
NPT Coupling Body (Non-Valved)
NPT Valved Coupling Body
Unit Size:
Each
Product Code. 30054082
Brand: CPC (Colder) 5011228
Only null left
Add to basket
Add to basket
Make sterile connections quick and easy with the push-to-connect fittings
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Connect to a female coupling body for easy fluid transfer. Audible click provides added assurance of a secure connection. Quick one-step disconnection process minimize user error
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/4 in.
Min. Operating Temperature (Metric) -40°F
Outlet Port Quick Disconnect Insert x Hose Barb
Shape Straight
Color Amber tint
Product Type NPT Valved Coupling Body