Learn More
CPC (Colder) HFC Quick-Disconnect Fittings, Hose Barb Insert, Polysulfone with Alloy C-276 Valve
Product Code. 30054205
Click to view available options
Diameter (English) Inner:
1/2 in.
1/4 in.
3/8 in.
Unit Size:
Each
Product Code. 30054205
Brand: CPC (Colder) 5011231
Valved connectors ensure sterility is maintained on both sides of the system during the disconnection process
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Connect to a female coupling body for easy fluid transfer. Audible click provides added assurance of a secure connection. Quick one-step disconnection process minimize user error. Alloy C-276 valve spring stops the flow and offers broad application compatibility. Laser-etched item and lot numbers offer batch traceability
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 3/8 in.
Material Polysulfone
Outlet Port Quick Disconnect Insert x Hose Barb
Sterilization Method 132°C
Shape Straight
Material (Seal) Silicone