missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoic-5,5,6,6,6-d5 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Hexanoic Acid-d5 |
| CAS | 123167-40-8 |
| Molecular Formula | C6H7D5O2 |
| Quantity | 0.25 g |
| Synonym | Hexanoic-5,5,6,6,6-d5 Acid |
| InChI Formula | InChI=1 S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i1D3,2D2 |
| SMILES | OC(CCCC([2 H])([2 H])C([2 H])([2 H])[2 H])=O |
| IUPAC Name | hexanoic-5,5,6,6,6-d5 acid |
| Molecular Weight (g/mol) | 121.19 |
| Formula Weight | 121.12 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction