missing translation for 'onlineSavingsMsg'
Learn More

Hexahydrophenyl Cinacalcet, TRC

Product Code. 30134627
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134627 250 mg 250mg
30003706 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134627 Supplier Toronto Research Chemicals Supplier No. TRCH291100250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271930-12-1
Molecular Formula C22H28F3N
Molecular Weight (g/mol) 363.46
Synonym (alphaR)-alpha-Methyl-N-[3-[3-(trifluoromethyl)cyclohexyl]propyl]-1-naphthalenemethanamine, N-((R)-1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)cyclohexyl)propan-1-amine, Cinacalcet Impurity F
IUPAC Name N-[(1R)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)cyclohexyl]propan-1-amine
SMILES C[C@@H](NCCCC1CCCC(C1)C(F)(F)F)c2cccc3ccccc23

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H28F3N/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25/h2-3,5,9-10,12-13,16-17,19,26H,4,6-8,11,14-15H2,1H3/t16-,17?,19?/m1/s1
Formula Weight 363.2174
Chemical Name or Material Hexahydrophenyl Cinacalcet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.