Learn More
Hexahydro-[1,4]dioxino[2,3-b][1,4]dioxine-2,3,6,7-tetrol, TRC
Specifications
Specifications
| Chemical Name or Material | Hexahydro-[1,4]dioxino[2,3-b][1,4]dioxine-2,3,6,7-tetrol |
| CAS | 4405-13-4 |
| Molecular Formula | C6H10O8 |
| Quantity | 1 g |
| Synonym | Glyoxal Trimer Dihydrate, 2,3,4a,6,7,8a-Hexahydro-[1,4]dioxino[2,3-b][1,4]dioxine-2,3,6,7-tetrol |
| InChI Formula | InChI=1S/C6H10O8/c7-1-2(8)12-6-5(11-1)13-3(9)4(10)14-6/h1-10H |
| SMILES | OC1OC2OC(O)C(O)OC2OC1O |
| IUPAC Name | 2,3,4a,6,7,8a-hexahydro-[1,4]dioxino[2,3-b][1,4]dioxine-2,3,6,7-tetrol |
| Molecular Weight (g/mol) | 210.13 |
| Formula Weight | 210.0376 |