missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Bromide, TRC

Product Code. 17965065
Change view
Click to view available options
Quantity:
50 mg
50 g
100 g
Unit Size:
100g
50g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17965065 50 g 50g
17963985 50 mg 50mg
17930415 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17965065 Supplier Toronto Research Chemicals Supplier No. TRCH29386050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19 H42 N . Br
Molecular Weight (g/mol) 364.45
Synonym 1-Hexadecanaminium, N,N,N-trimethyl-, bromide (1:1), 1-Hexadecanaminium, N,N,N-trimethyl-, bromide (9CI), Ammonium, hexadecyltrimethyl-, bromide (8CI), Hexadecyltrimethylammonium bromide (6CI), 1-Hexadecyltrimethylammonium bromide, Accelerator 1631, Accelerator DT 3126-1, Acetoquat CTAB, Arquad 16/60, BCTA, Bromat, C16TAB, CTAB, CTABr, CTMAB, CTMB, Catinal HTB 70ET, Cetremide, Cetrimonium bromide, Cetyltrimethylammonium bromide, Cirrasol OD, HDTMA-Br, HTAB, Hexadecanyltrimethylammonium bromide, Hexadecyltrimethylamine bromide, Lauroseptol, Lissolamine, Lissolamine A, Micol, N,N,N-Trimethyl-1-hexadecanaminium bromide, N,N,N-Trimethylhexadecan-1-ammonium bromide, N,N,N-Trimethylhexadecylammonium bromide, N-Cetyl-N,N,N-trimethylammonium bromide, N-Cetyltrimethylammonium bromide, N-Hexadecyl-N,N,N-trimethylammonium bromide, Palmityltrimethylammonium bromide, Pollacid, Quamonium, Rhodaquat M 242B99, Softex KW, Trimethylcetylammonium bromide, Trimethylhexadecylammonium bromide, Varisoft CTB 40, ZYS 08, n-Hexadecyltrimethylammonium bromide
IUPAC Name hexadecyl(trimethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1
Formula Weight 363.2501
Chemical Name or Material Cetrimonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.