missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic-9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d17 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-D17 Acid |
| CAS | 81462-28-4 |
| Molecular Formula | C16 D17 H15 O2 |
| Quantity | 0.1 g |
| Synonym | Hexadecanoic-9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d17 acid (9 CI), Hexadecanoic-9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-D17 Acid, Palmitic acid-9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d17, [9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-2H17]Hexadecanoic acid, [9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-2H17]Palmitic acid, Hexadecanoic Acid-9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d17 |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCCCCCC(=O)O |
| IUPAC Name | 9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-heptadecadeuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 273.5288 |
| Formula Weight | 273.3469 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction