missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic-5,5,6,6-d4 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Palmitic Acid-d4 |
| CAS | 75736-47-9 |
| Molecular Formula | C16 D4 H28 O2 |
| Quantity | 0.1 g |
| Synonym | Hexadecanoic-5,5,6,6-d4 acid (9 CI), Hexadecanoic-5,5,6,6-D4 acid, [5,5,6,6-2H4]Hexadecanoic acid, Hexadecanoic acid-5,5,6,6-d4, Palmitic-5,5,6,6-d4 acid, Palmitic acid-5,5,6,6-d4 |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i11D2,12D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCC)C([2 H])([2 H])CCCC(=O)O |
| IUPAC Name | 5,5,6,6-tetradeuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 260.449 |
| Formula Weight | 260.265 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction