missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic-13,13-d2 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-13,13-D2 Acid |
| CAS | 62690-28-2 |
| Molecular Formula | C16 D2 H30 O2 |
| Quantity | 0.25 g |
| Synonym | Hexadecanoic-13,13-d2 acid (9 CI), Hexadecanoic-13,13-D2 Acid, Palmitic acid-13,13-d2, [13,13-2H2]Hexadecanoic acid, [13,13-2H2]Palmitic acid, Hexadecanoic Acid-13,13-d2 |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i4D2 |
| SMILES | [2 H]C([2 H])(CCC)CCCCCCCCCCCC(=O)O |
| IUPAC Name | 13,13-dideuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 258.4364 |
| Formula Weight | 258.2528 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction