missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexacosanoic acid, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 87.0°C to 89.0°C |
| Color | White |
| Density | 0.8100g/mL |
| Boiling Point | 250.0°C |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Linear Formula | CH3(CH2)24COOH |
| Refractive Index | 1.4301 |
| Mehr anzeigen |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur