missing translation for 'onlineSavingsMsg'
Learn More

Hetastarch, Technical Grade, TRC

Product Code. 30138530
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138530 10 g 10g
30133568 50 g 50g
30008454 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138530 Supplier Toronto Research Chemicals Supplier No. TRCH28955010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-27-0
Molecular Formula C22H44O17
Molecular Weight (g/mol) 580.6
Synonym 2-Hydroxyethyl Starch, 2-Hydroxyethyl Starch Ether, Amaizo 742D, Coatmaster K 54F, Coatmaster K 550, Elohast, Elohes, Essex 1360, Essex Gum 1360, Hespan, Hespander, Hesteril, PG 280, PG 295, PG 380, Pen-Cote, PenFilm HV 421182, enford Gum 280, Penford Gum 290, Penford Gum 295, Tapioca Starch Hydroxyethyl ether, Tetraspan, Unique Gum 3010, Venofundin, Volekam, Volex, Voluven;
SMILES C(COC[C@@H]1[C@H]([C@@H]([C@H]([C@H](O1)OCCO)OCCO)OCCO)OCCO)O.C([C@@H]1[C@H]([C@@H]([C@H]([C@H](O1)O)O)O)O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H32O11.C6H12O6/c17-1-6-22-11-12-13(23-7-2-18)14(24-8-3-19)15(25-9-4-20)16(27-12)26-10-5-21;7-1-2-3(8)4(9)5(10)6(11)12-2/h12-21H,1-11H2;2-11H,1H2/t12-,13-,14+,15-,16+;2-,3-,4+,5-,6+/m11/s1
Chemical Name or Material Hydroxyethyl starch
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.