Learn More
Thermo Scientific™ Heratherm™ Advanced Protocol Security Ovens
Product Code. 12858231 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
61 L
62 L
97 L
99 L
168 L
170 L
Type:
Gravity Convection
Mechanical Convection Oven
Electrical Requirements:
208/240 V 60 Hz
230 V 50/60 Hz
Material:
Coated Steel
Stainless Steel
Unit Size:
Each
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 12858231
Brand: Thermo Scientific™ 51028128
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Ensure optimal sample protection with these advanced protocol security ovens that feature an extra layer of security for applications where process reliability and security are paramount.
Protect samples effectively with Thermo Scientific™ Heratherm™ Advanced Protocol Security Ovens, which feature an extra layer of security for applications where process reliability and security are paramount. A standard over-temperature alarm and an additional under-temperature alarm ensure sample stability. A door lock prevents disruption or tampering of samples, while a door alarm notifies the operator when the door is left open accidentally. Choose a stainless steel exterior for demanding pharmaceutical and clinical applications.
Features:
  • Broad temperatures range — up to 330°C
  • Advanced digital timer for daily or weekly on/off cycles
  • Stainless-steel interior is easy to clean and corrosion resistant (steel quality AISI 304)
  • Programmable controller for temperature ramps and dwells
  • Up to 10 programs can be saved, each with 10 discrete steps
  • Electronically controlled fan speed and damper position can be incorporated
  • Programs can be repeated automatically
  • Access port allows the introduction of sensors for independent data monitoring
  • Simple calibration routine ensures temperature accuracy over time
  • Boost function enables rapid heating — no need to operate the oven 24/7
Ordering Information:
Availability may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Controller Type Programmable Microprocessor control with vacuum fluorescent display
Capacity (Metric) 97 L
Temperature (Metric) 50°C to 330°C
Type Mechanical Convection Oven
Material Coated Steel
No. of Shelves 2 supplied/16 max.
Plug Type Nema 6-20
Temperature Stability ±0.2°C
Timer Weekly / Real time / Hour
Dimensions (D x W x H) Interior 13.5 x 17.2 x 23.9 in. (34.3 x 43.8 x 60.8 cm)
Dimensions (L x W x H) 22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Length (English) 22.2 in.
Length (Metric) 56.5 cm
Width (English) 25.2 in.
Width (English) Interior 17.2 in.
Width (Metric) 64 cm
Width (Metric) Interior 43.8 cm
Height (English) 32.3 in.
Height (English) Interior 23.9 in.
Height (Metric) 82 cm
Height (Metric) Interior 60.8 cm
Depth (English) Interior 13.5 in.
Depth (Metric) Interior 34.3 cm
Electrical Requirements 208/240 V 60 Hz
Voltage 208/240 V
Wattage 3360 w
Amperage 14 A
Frequency 60 Hz
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.