Learn More
Heptanoic anhydride, 99%, Thermo Scientific Chemicals
Catalog Number 10644692
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 626-27-7
Melting Point -12°C
Color Yellow
Density 0.9170g/mL
Boiling Point 278°C to 282°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C14H26O3
Linear Formula [CH3(CH2)5CO]2O
Refractive Index 1.4320 to 1.434
MDL Number MFCD00009536
Quantity 50 g
Beilstein 02, 340
Specific Gravity 0.917
Synonym heptanoic anhydride, enanthic anhydride, heptanoic acid, anhydride, heptanoyl anhydride, n-heptanoic acid anhydride, n-heptanoic anhydride, heptanoic acid, 1,1'-anhydride, oenanthic anhydride, bisenanthic anhydride, enanthic acid anhydride
InChI Key DAPZDAPTZFJZTO-UHFFFAOYSA-N
SMILES CCCCCCC(=O)OC(=O)CCCCCC
IUPAC Name heptanoyl heptanoate
Molecular Weight (g/mol) 242.36
PubChem CID 69376
Formula Weight 242.36
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Heptanoic anhydride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Heptanoic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.