Learn More
Heptanoic-5,5,6,6-d4 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Heptanoic Acid-5,5,6,6-D4 |
| CAS | 352431-36-8 |
| Molecular Formula | C7 D4 H10 O2 |
| Quantity | 0.05 g |
| Synonym | Heptanoic-5,5,6,6-d4 acid (9 CI), Heptanoic Acid-5,5,6,6-D4, Heptanoic acid-d4, Enanthic acid-d4 |
| InChI Formula | InChI=1 S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(C)C([2 H])([2 H])CCCC(=O)O |
| IUPAC Name | 5,5,6,6-tetradeuterioheptanoic acid |
| Molecular Weight (g/mol) | 134.2095 |
| Formula Weight | 134.1245 g/mol |
| Show More |