Learn More
Heptanal, 97%
Product Code. 11369819
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL
Unit Size:
1000mL
100mL
250mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369819 250 mL 250mL
11389819 100 mL 100mL
11399819 500 mL 500mL
11379819 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11369819 Supplier Thermo Scientific Alfa Aesar Supplier No. B23830.AK
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
CAS: 111-71-7 | C7H14O | 114.19 g/mol
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
Solubility
Soluble in water 1.3 g/L.
Notes
Hygroscopic. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-71-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00007028
InChI Key FXHGMKSSBGDXIY-UHFFFAOYSA-N
Synonym heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
PubChem CID 8130
ChEBI CHEBI:34787
IUPAC Name heptanal
SMILES CCCCCCC=O
Specifications
Melting Point °C to 43°C
Density 0.818
Boiling Point 152°C to 154°C
Flash Point 35°C (95°F)
Assay Percent Range 0.97
Refractive Index 1.4125
Quantity 250 mL
UN Number UN3056
Beilstein 1560236
Sensitivity Air Sensitive
Merck Index 14,4658
Solubility Information Soluble in water 1.3g/L.
Formula Weight 114.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Heptanal