Learn More
Heptaminol, TRC
Specifications
Specifications
| Chemical Name or Material | Heptaminol |
| CAS | 372-66-7 |
| Molecular Formula | C8 H19 N O |
| Quantity | 100 mg |
| Synonym | 6-Amino-2-methyl-2-heptanol, (5-Hydroxy-1,5-dimethylhexyl)amine, 2-Amino-6-hydroxy-6-methylheptane, 2-Methyl-6-amino-2-heptanol, 6-Hydroxy-6-methyl-2-aminoheptane, 6-Hydroxy-6-methyl-2-heptamine, 6-Hydroxy-6-methyl-2-heptylamine, 6-Methyl-2-amino-6-heptanol, Corasore, Ginkor Fort, Heptaminol, 6-Amino-2-methyl-2-heptanol |
| InChI Formula | InChI=1S/C8H19NO/c1-7(9)5-4-6-8(2,3)10/h7,10H,4-6,9H2,1-3H3 |
| SMILES | CC(N)CCCC(C)(C)O |
| IUPAC Name | 6-amino-2-methylheptan-2-ol |
| Molecular Weight (g/mol) | 145.24 |
| Formula Weight | 145.1467 |