missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptafluoro-1-methoxypropane, TRC
Specifications
Specifications
| Chemical Name or Material | Heptafluoro-1-methoxypropane |
| CAS | 375-03-1 |
| Molecular Formula | C4H3F7O |
| Quantity | 5 g |
| Synonym | 1,1,1,2,2,3,3-Heptafluoro-3-methoxypropane, 1,1,2,2,3,3,3-Heptafluoro-1-methoxypropane, 1-Methoxy-1,1,2,2,3,3,3-heptafluoropropane, 1-Methoxyheptafluoropropane, 1-Methoxyperfluoropropane, E 347, HFE 301, HFE 347, HFE 347MCC1, HFE 347mcc, HFE 7000, Heptafluoropropyl Methyl Ether, Methyl Heptafluoropropyl Ether, Methyl Perfluoropropyl Ether, Novec 7000, Novec HFE 7000, Perfluoropropyl Methyl Ether, R-E 347mcc, 1RE 347mcc |
| InChI Formula | InChI=1S/C4H3F7O/c1-12-4(10,11)2(5,6)3(7,8)9/h1H3 |
| SMILES | COC(C(C(F)(F)F)(F)F)(F)F |
| IUPAC Name | 1,1,1,2,2,3,3-Heptafluoro-3-methoxypropane;1,1,2,2,3,3,3-Heptafluoro-1-methoxypropane; 1-Methoxy-1,1,2,2,3,3,3-heptafluoropropane; 1-Methoxyheptafluoropropane; 1-Methoxyperfluoropropane; E 347; HFE 301;HFE 347;HFE 347MCC1; HFE 347mcc; HFE 7000; Heptafluoropropyl Methyl Ether; Methyl Heptafluoropropyl Ether; Methyl Perfluoropropyl Ether; Novec 7000; Novec HFE 7000; Perfluoropropyl Methyl Ether; R-E 347mcc; 1RE 347mcc; |
| Molecular Weight (g/mol) | 200.06 |
| Formula Weight | 200.01 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?