Learn More
STEMCELL Technologies HepatiCult™ Organoid Growth Medium (Mouse)
Product Code. 17129231
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17129231 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17129231 Supplier STEMCELL Technologies Supplier No. 06030
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Cell culture medium for establishment and maintenance of mouse hepatic progenitor organoids
  • HepatiCult™ Organoid Growth Medium (Mouse) is a serum-free, defined cell culture medium for establishment and maintenance of mouse hepatic progenitor organoids
  • These organoids, or 'mini-livers', provide an in vitro organotypic culture system for studying hepatic stem and progenitor cells
  • Organoids grown in HepatiCult™ feature an epithelium expressing genes marking hepatic stem and progenitor cells (PROM1, AXIN2, SOX9 and CD44), ducts (KRT19 and HNF1b) and hepatocytes (HNF4a, AFP)
  • Hepatic organoids can be passaged every 4 - 7 days, can be cryopreserved, and are primed for downstream differentiation
  • HepatiCult™ supports mouse hepatic organoid culture either embedded in Corning™ Matrigel™ domes or in a dilute Matrigel™ suspension
  • Organoid culture enables convenient in vitro characterization of the hepatic epithelium in a physiologically relevant system and reduces the need for animal use
Specifications
Cell Type Hepatocyte Cells
For Use With (Application) Cell Culture; Expansion; Maintenance; Organoid Culture
Quantity 1 Kit