Learn More
ACROBiosystems HEK293/Human TrkB Stable Cell Line
Product Code. 31451815
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31451815 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31451815 Supplier ACROBiosystems Supplier No. CHEKATP0822Vials
This gene encodes a member of the neurotrophic tyrosine receptor kinase(NTRK)family.This kinase is a membrane-bound receptor that upon neurotrophin binding,phosphorylates itself and members of the MAPK pathway.Signalling through this kinase leads to cell differentiation.Mutations in this gene have been associated with obesity and mood disorders.
This gene encodes a member of the neurotrophic tyrosine receptor kinase(NTRK)family.This kinase is a membrane-bound receptor that upon neurotrophin binding,phosphorylates itself and members of the MAPK pathway.Signalling through this kinase leads to cell differentiation.Mutations in this gene have been associated with obesity and mood disorders.
Specifications
Quantity 2 Vials