Learn More
ACROBiosystems HEK293/Human Transferrin R Stable Cell Line
Product Code. 31451823
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31451823 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31451823 Supplier ACROBiosystems Supplier No. CHEKATP0892Vials
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia and lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia and lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.
Specifications
Quantity 2 Vials