Learn More
ACROBiosystems HEK293/Human Claudin-18.2 Stable Cell Line
Product Code. 31453657
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31453657 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31453657 Supplier ACROBiosystems Supplier No. CHEKATP0332Vials
Claudin-18.2 is a member of the tight junction protein family. Under physiological conditions, it is only expressed at low levels in the differentiated epithelial cells of the gastric mucosa; whereas under pathological conditions, it exhibits abnormally high expression in various tumors such as gastric cancer and pancreatic cancer, making it an ideal biomarker for tumor-targeted therapy.
Claudin-18.2 is a member of the tight junction protein family. Under physiological conditions, it is only expressed at low levels in the differentiated epithelial cells of the gastric mucosa; whereas under pathological conditions, it exhibits abnormally high expression in various tumors such as gastric cancer and pancreatic cancer, making it an ideal biomarker for tumor-targeted therapy.
Specifications
Quantity 2 Vials