Learn More
ACROBiosystems HEK293/Human Anti-CD19 Stable Cell Line
Product Code. 31453356
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31453356 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31453356 Supplier ACROBiosystems Supplier No. CHEKATS0562Vials
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Chimeric antigen receptors (CARs) comprise interact domains, of which target cancer cell expressed antigens and allow T cell to efficiently kill the cancer cell. FMC63 is an IgG2a mouse monoclonal antibody specific for CD19, which is a target for the immunotherapy of B lineage leukaemias and lymphomas.
Chimeric antigen receptors (CARs) comprise interact domains, of which target cancer cell expressed antigens and allow T cell to efficiently kill the cancer cell. FMC63 is an IgG2a mouse monoclonal antibody specific for CD19, which is a target for the immunotherapy of B lineage leukaemias and lymphomas. FMC63 scFv is the most commonly used ectodomain component of CD19-specific CARs. Our CAR-like cells apply the HEK293 cells for the expression of FMC63 which will be presented on the cell membrane. It mimics the real CAR-T cell reaction and it is used as positive control for Anti-CD19 scFv Antibody flow cytometry detection.
Specifications
Quantity 2 Vials