Learn More
Horst Heating cord, type HS 42, L = 1.3 m 120 W.
Brand:  Horst 23004
Heating cord, type HS 42, L = 1.3 m 120 W.
Product Code. 10047121
  • ONLINE EXCLUSIVE PRICE
    80.38€
    66.50€ / Each
    Save 13.88€
    17% Off
Only null left
Estimated Shipment: 07-01-2026