Learn More
Heidolph™ Hei-VAP Core HL G3 XL, EU Plug
Product Code. 17323290 Shop All Heidolph Instruments Products
Change view
Click to view available options
Capacity (Metric):
5 L Evaporating Flask, 4.5 L Bath
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17323290 5 L Evaporating Flask, 4.5 L Bath Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17323290 Supplier Heidolph™ Supplier No. 5710130500
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 28-12-2026
The Hei-VAP Core HL G3 XL is focused on the essentials: a clear digital display with access to the basic parameters at any time.
  • Clear digital display showing actual and set values
  • Two separate knobs with dynamic control for setting the rotation speed and heating bath temperature: fast turning for large jumps, slow turning for precise setting of the set values
  • Accidental changing of the values can be prevented by the lock function
  • Intuitive LED ring light system makes activated functions clearly visible even from a distance
  • Easy adjustment of the immersion depth up to 155 mm and the inclination angle from 20° to 80°
  • Standby function and residual heat warning at heating bath temperatures above 50°C
  • Hand lift model with G3 XL vertical glassware - achieve higher evaporation rates due to expanded cooling surface of 2,200 cm2
  • The standby button stops all functions.
TRUSTED_SUSTAINABILITY
Specifications
Weight (Metric) 15 kg
Weight (English) 33.1 lb.
Capacity (Metric) 5 L Evaporating Flask, 4.5 L Bath
Speed 10 to 280 rpm
Temperature Range (Metric) 20°C to 210°C
Display LCD
Motor Type Brushless DC Motor
Temperature Range (English) 68°F to 410°F
Wattage 1400 W
Glassware Assembly Type G3 XL Vertical Glassware
IP Rating IP20
Material Stainless Steel
Controller Type Electronic
Temperature Accuracy ±1 K
Angle 20° to 80°
Type Hand Lift Rotary Evaporator
Plug Type EU Plug