Learn More
Heidolph™ Hei-Connect PT1000 Clamping System for Bath Attachments
Shop All Heidolph Instruments Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
3 to 5 L Heat-On Blocks
missing translation for 'unitSize'
Each
Description
- Includes support rod and attachment with cable inlet
- Provides maximum safety and stability in its vertical alignment
Specifications
Specifications
| For Use With (Equipment) | 3 to 5 L Heat-On Blocks |
| For Use With (Application) | Vertical Support for Bath Attachments |
| Type | Support Stand |
| Material | Stainless Steel |