Learn More
Heidolph™ Hei-CHILL 400 Pro Circulating Chiller
Product Code. 17987140 Shop All Heidolph Instruments Products
Click to view available options
Plug Type:
EU Plug
Unit Size:
Each
Product Code. 17987140
Brand: Heidolph™ 5910164000
This space-saving Circulating Chiller with a cooling capacity of 400 watt is ideal for a large number of evaporation tasks in the laboratory
  • Compact light weight for a flexible positioning on the laboratory bench, in a fume hood or, due to its increased pumping capacity, also under the laboratory bench
  • Sustainable and environmentally friendly using natural refrigerants (R-290) and an optimized control
  • RS-232 interface for activating your device via a Hei-VAP Expert Control or Ultimate Control or via the Heidolph laboratory software solutions
  • Low operating noises for a trouble-free work
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1.5 to 4.0 L
Condenser Type Air-cooled
Cooling Capacity 400 W at 20°C
Flow Rate 6 L/min.
For Use With (Equipment) Rotary Evaporators
Temperature Range -10°C up to 40°C
Type Recirculating Chiller
Plug Type EU Plug
Electrical Requirements 230 V/50 Hz
Voltage 230 V