Learn More
Rogo Sampaic™ Heavy Crystallizing Dishes
Ideal for crystallization needs in the laboratory. Rogo Sampaic™ Heavy Crystallizing Dishes are manufactured from clear soda lime glass.
Brand:  Rogo Sampaic™ CNN026
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11702704
  • 213.06€ / Pack of 3
Estimated Shipment: 05-06-2024
Description
  • Also excellent for storage
  • Available in a range of sizes
Specifications
Specifications
2 L
Crystallizing Dish
96 mm
205 mm