Learn More
Horst™ Heating Tape Outer Stainless Steel Braid
Product Code. 10178541
Change view
Click to view available options
Length (Metric):
1.5 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10178541 1.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10178541 Supplier Horst™ Supplier No. 20333
  • ONLINE EXCLUSIVE PRICE
    244.90€
    180.00€ / Pack of 1
    Save 64.90€
    26% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Multiple electrical glass-yarn insulation and protective braid.
  • Multiple electrical glass-yarn insulation.
  • Protective outer braid.
Specifications
Length (Metric) 1.5 m
Liner Material Glass Yarn, Stainless Steel
Temperature (Metric) 350°C (Nominal)