Learn More
Horst™ Heating Hose
Product Code. 12684546
Change view
Click to view available options
Diameter (Metric):
4mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12684546 4mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12684546 Supplier Horst™ Supplier No. 822637
  • ONLINE EXCLUSIVE PRICE
    958.65€
    635.12€ / Pack of 1
    Save 323.53€
    33% Off
Estimated Shipment: 19-10-2026
Specifications
Diameter (Metric) 4mm
Length (Metric) 1500 mm
Product Type Heating Hose
Temperature (Metric) 200°C