Learn More
BRAND™ Heat Sealing Film
Product Code. p-200104831 Shop All Brand Products
Click to view available options
Temperature Range (Metric):
-20°C to +120°C
-200°C to +110°C
-80°C to +110°C
Unit Size:
Pack of 100
Product Code. 17827777
Brand: BRAND™ 701430
Only null left
Add to basket
Add to basket
Heat sealing technology for effective and durable plate sealing.
Heat sealing films provide very good protection against evaporation and contamination due to the tight seal.
Aluminum foils are used for PP and PS plates. They are resistant to solvents and are used both at low temperatures and for storage at room temperature. Depending on the application, they are peelable or pierceable.
The transparent film in PE is used for PP sheets. Due to its good optical properties it is suitable for qPCR and colorimetric assays.
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Material Aluminum
Sterility Non-sterile
Autoclavable Not Autoclavable
Adhesive Aluminum Heat Seal
Pierceable Yes
Temperature Range (Metric) -20°C to +120°C
Fit for Purpose PCR Ready