Description
- Heat-On inserts are suitable for two 25 mL, 50 mL, or 100 mL round bottom flasks
- Multi-Well holder can enclose two Heat-On inserts at the same time, allowing for multiple combinations
- In order to measure precisely with a temperature sensor, a small hole is provided on one side of the Heat-On Block
- Scratch and chemical-resistant Multi-Well holder
- Safety lifting handles are optional and available separately
Specifications
Specifications
| For Use With (Equipment) | Round Bottom Flasks |
| For Use With (Application) | Heating Glassware |
| Includes | Holder and 6 Inserts |
| Temperature (Metric) | 260°C |
| Coating | PTFE |
| Material | Aluminum |