Learn More
Huenersdorff™ HDPE Universal Container
Click to view available options
Capacity (Metric):
200 mL
400 mL
700 mL
1000 mL
2000 mL
Height (Metric):
42 mm
55 mm
64 mm
88 mm
110 mm
127 mm
Unit Size:
Pack of 16
Pack of 20
Pack of 25
Pack of 30
Description
- Universal container
- Natural for varied applications
- Made of food-safe polyethylene
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Closure Material | LDPE |
| Closure Type | Hooded Lid |
| Color | Natural |
| Material | Polyethylene |
| Shape | Square |
| For Use With (Application) | Food Safe |
| For Use With (Equipment) | Deep Freezer, Dishwasher, Microwave |
| Includes | Tightly Closing LDPE Lid |
| Mounting Type | Stackable |
| Show More |