Learn More
BRAND™ HDPE Straight Tubing Adapter
Shop All Brand Products
Click to view available options
Tubing Size:
11 to 14mm I.D.
18 to 20.5mm I.D
Unit Size:
Pack of 10
Description
- High-density Polyethylene construction
- Straight tubing adapter includes a large and small end to accommodate many sizes.
- Hose barbs provide a secure fit.
Specifications
Specifications
| Material | HDPE |
| Overall Length (Metric) | 66 mm |
| For Use With | 8 to 10/11-14mm tubing I.D. |
| Packaging Quantity | 10 Pack |
| Tubing Size | 11 to 14mm I.D. |
| Shape | Straight |
| Product Type | Tubing Adapter |