Learn More
Huenersdorf™ HDPE Specimen Containers
Product Code. 11756735
Change view
Click to view available options
Capacity (Metric):
500 mL
2000 mL
Shape:
Round
Square
Unit Size:
Pack of 20
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Shape Unit Size
11756735 2000 mL Square Pack of 20
11746745 2000 mL Round Pack of 25
11716735 500 mL Square Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11756735 Supplier Huenersdorf™ Supplier No. 910500
  • ONLINE EXCLUSIVE PRICE
    233.10€
    83.23€ / Pack of 20
    Save 149.87€
    64% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Secure your items securely in Heunersdorf™ HDPE Specimen Containers, which are made of food-safe polypropylene (PP) for assorted uses.
  • Resistant to acids and chemicals
  • Tightly closing lid
  • Deep freezable
Specifications
Capacity (Metric) 2000 mL
Closure Color Natural
Closure Material LDPE
Closure Type Hooded Lid
Color Natural
Material Polyethylene
Shape Square
For Use With (Application) Food Safe
For Use With (Equipment) Deep Freezer, Dishwasher, Microwave
Includes Closure
Mounting Type Stackable
Product Type Specimen Container
Quantity 20 Pack
Length (Metric) 103 mm
Width (Metric) 255 mm
Height (Metric) 94 mm