Learn More
Brand™ HDPE Scoops
Shop All Brand Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Length (Metric):
350 mm
400 mm
Unit Size:
Pack of 6
Description
- Manufactured from HDPE
- Natural colored
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Color | Natural |
| Product Type | Scoop |
| Material | HDPE |
| Length (Metric) | 400 mm |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur